missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pentanone 98.0+%, TCI America™

$67.11 - $251.65

Chemical Identifiers

CAS 565-61-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00009336
InChI Key UIHCLUNTQKBZGK-UHFFFAOYSA-N
Synonym 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
PubChem CID 11262
IUPAC Name 3-methylpentan-2-one
SMILES CCC(C)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M03885ML
View Documents
TCI America
M03885ML
5 mL
Each for $67.11
Only null left
 
M038825ML
View Documents
TCI America
M038825ML
25 mL
Each for $251.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

565-61-7
100.161
UIHCLUNTQKBZGK-UHFFFAOYSA-N
11262
CCC(C)C(=O)C
C6H12O
MFCD00009336
3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
3-methylpentan-2-one

Specifications

565-61-7
116°C
MFCD00009336
1224
UIHCLUNTQKBZGK-UHFFFAOYSA-N
3-methylpentan-2-one
11262
≥98.0% (GC)
3-Methyl-2-pentanone
Yellow
C6H12O
5 mL
3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
CCC(C)C(=O)C
100.161
100.16
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0542

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'