Learn More
3-Methyl-3-heptanol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2095914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Methyl-3-heptanol | |
0.83 | |
58°C (136°F) | |
MFCD00021841 | |
UN1987 | |
PQOSNJHBSNZITJ-UHFFFAOYSA-N | |
3-methylheptan-3-ol | |
11710 | |
98% |
5582-82-1 | |
64°C (201 mmHg) | |
C8H18O | |
25 g | |
3-methyl-3-heptanol, 3-heptanol, 3-methyl, 2-ethyl-2-hexanol, butylethylmethylcarbinol, 3-heptanol, 3-methyl-8ci 9ci, acmc-209lpn | |
CCCCC(C)(CC)O | |
130.231 | |
130.23 |
Chemical Identifiers
5582-82-1 | |
130.231 | |
PQOSNJHBSNZITJ-UHFFFAOYSA-N | |
11710 | |
CCCCC(C)(CC)O |
C8H18O | |
MFCD00021841 | |
3-methyl-3-heptanol, 3-heptanol, 3-methyl, 2-ethyl-2-hexanol, butylethylmethylcarbinol, 3-heptanol, 3-methyl-8ci 9ci, acmc-209lpn | |
3-methylheptan-3-ol |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA : No
RUO – Research Use Only