Learn More
3-Methyl-3-hexanol, 99%, Thermo Scientific Chemicals
$148.10 - $507.67
Chemical Identifiers
CAS | 597-96-6 |
---|---|
Molecular Formula | C7H16O |
Molecular Weight (g/mol) | 116.204 |
MDL Number | MFCD00021839 |
InChI Key | KYWJZCSJMOILIZ-UHFFFAOYSA-N |
Synonym | 3-methyl-3-hexanol, 3-hexanol, 3-methyl, 2-ethyl-2-pentanol, 3-methyl-hexanol-3, 3-methyl-hexanol-3 german, acmc-20an0r, 4-01-00-01749 beilstein handbook reference |
PubChem CID | 11708 |
IUPAC Name | 3-methylhexan-3-ol |
SMILES | CCCC(C)(CC)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0462506
|
Thermo Scientific Chemicals
L0462506 |
5 g |
Each for $148.10
|
|
||||||
AAL0462514
|
Thermo Scientific Chemicals
L0462514 |
25 g |
Each for $507.67
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
597-96-6 | |
116.204 | |
KYWJZCSJMOILIZ-UHFFFAOYSA-N | |
11708 | |
CCCC(C)(CC)O |
C7H16O | |
MFCD00021839 | |
3-methyl-3-hexanol, 3-hexanol, 3-methyl, 2-ethyl-2-pentanol, 3-methyl-hexanol-3, 3-methyl-hexanol-3 german, acmc-20an0r, 4-01-00-01749 beilstein handbook reference | |
3-methylhexan-3-ol |
Specifications
597-96-6 | |
143°C | |
C7H16O | |
MFCD00021839 | |
UN1987 | |
3-methyl-3-hexanol, 3-hexanol, 3-methyl, 2-ethyl-2-pentanol, 3-methyl-hexanol-3, 3-methyl-hexanol-3 german, acmc-20an0r, 4-01-00-01749 beilstein handbook reference | |
CCCC(C)(CC)O | |
116.204 | |
116.2 | |
3-Methyl-3-hexanol |
0.823 | |
45°C (113°F) | |
1.4235 | |
5 g | |
1719032 | |
KYWJZCSJMOILIZ-UHFFFAOYSA-N | |
3-methylhexan-3-ol | |
11708 | |
99% |
Safety and Handling
GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 209-910-4
RTECSNumber : MP0950000
TSCA : Yes
RUO – Research Use Only