Learn More
3-Methyl-3-penten-2-one, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 438440100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Methyl-3-penten-2-one | |
97.5 | |
0.8750g/mL | |
34°C | |
97.5% min. (GC) | |
C6H10O | |
10g | |
3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene | |
ZAMCMCQRTZKGDX-SNAWJCMRSA-N | |
(E)-3-methylpent-3-en-2-one | |
5364579 | |
98% |
565-62-8 | |
100.0 | |
136°C to 139°C | |
Authentic | |
Glass bottle | |
CH3CH=C(CH3)COCH3 | |
0.875 | |
Solubility in water: 29.1g/L (20°C) | |
CC=C(C)C(=O)C | |
98.14 | |
98.14 |
Chemical Identifiers
565-62-8 | |
98.14 | |
3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene | |
(E)-3-methylpent-3-en-2-one |
C6H10O | |
ZAMCMCQRTZKGDX-SNAWJCMRSA-N | |
5364579 | |
CC=C(C)C(=O)C |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a P
GHS Signal Word: Warning
EINECSNumber : 209-283-7
RUO â Research Use Only