missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-penten-2-one 95.0+%, TCI America™

$156.59 - $661.75

Chemical Identifiers

CAS 565-62-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00151839
InChI Key ZAMCMCQRTZKGDX-SNAWJCMRSA-N
Synonym 3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
PubChem CID 5364579
IUPAC Name (E)-3-methylpent-3-en-2-one
SMILES CC=C(C)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M113925ML
View Documents
TCI America
M113925ML
25 mL
Each for $156.59
Only null left
 
M1139500ML
View Documents
TCI America
M1139500ML
500 mL
Each for $661.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

565-62-8
98.145
ZAMCMCQRTZKGDX-SNAWJCMRSA-N
5364579
CC=C(C)C(=O)C
C6H10O
MFCD00151839
3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
(E)-3-methylpent-3-en-2-one

Specifications

565-62-8
140°C
MFCD00151839
1224
ZAMCMCQRTZKGDX-SNAWJCMRSA-N
(E)-3-methylpent-3-en-2-one
5364579
≥95.0% (GC)
3-Methyl-3-penten-2-one
Yellow
C6H10O
25 mL
3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
CC=C(C)C(=O)C
98.145
98.15
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'