missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-4-nitrophenol, 98%, Thermo Scientific Chemicals

$80.18 - $191.75

Chemical Identifiers

CAS 2581-34-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007333
InChI Key PIIZYNQECPTVEO-UHFFFAOYSA-N
Synonym 4-nitro-m-cresol, 4-nitro-3-cresol, 4-nitro-3-methylphenol, 2-nitro-5-hydroxytoluene, 5-hydroxy-2-nitrotoluene, phenol, 3-methyl-4-nitro, 4-nitro-5-methylphenol, m-cresol, 4-nitro, p-nitro-m-cresol, 4-nitro-1-hydroxy-3-methylbenzene
PubChem CID 17412
ChEBI CHEBI:38683
IUPAC Name 3-methyl-4-nitrophenol
SMILES CC1=CC(O)=CC=C1[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0316314
View Documents
Thermo Scientific Chemicals
L0316314
25 g
Each for $80.18
 
AAL0316322
View Documents
Thermo Scientific Chemicals
L0316322
100 g
Each for $191.75
 
Description

Description

3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

Solubility
Soluble in water(1.34g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

2581-34-2
153.14
PIIZYNQECPTVEO-UHFFFAOYSA-N
17412
3-methyl-4-nitrophenol
C7H7NO3
MFCD00007333
4-nitro-m-cresol, 4-nitro-3-cresol, 4-nitro-3-methylphenol, 2-nitro-5-hydroxytoluene, 5-hydroxy-2-nitrotoluene, phenol, 3-methyl-4-nitro, 4-nitro-5-methylphenol, m-cresol, 4-nitro, p-nitro-m-cresol, 4-nitro-1-hydroxy-3-methylbenzene
CHEBI:38683
CC1=CC(O)=CC=C1[N+]([O-])=O

Specifications

2581-34-2
Phenol-like
MFCD00007333
1868105
Soluble in water(1.34g/L).
CC1=CC(O)=CC=C1[N+]([O-])=O
153.14
CHEBI:38683
98%
126°C to 130°C
C7H7NO3
25 g
4-nitro-m-cresol, 4-nitro-3-cresol, 4-nitro-3-methylphenol, 2-nitro-5-hydroxytoluene, 5-hydroxy-2-nitrotoluene, phenol, 3-methyl-4-nitro, 4-nitro-5-methylphenol, m-cresol, 4-nitro, p-nitro-m-cresol, 4-nitro-1-hydroxy-3-methylbenzene
PIIZYNQECPTVEO-UHFFFAOYSA-N
3-methyl-4-nitrophenol
17412
153.14
3-Methyl-4-nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H332-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 219-952-5

RTECSNumber : GP2625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only