missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals

$131.63 - $131.63

Chemical Identifiers

CAS 587-03-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004646
InChI Key JJCKHVUTVOPLBV-UHFFFAOYSA-N
Synonym 3-methylbenzyl alcohol, 3-methylphenyl methanol, m-tolyl carbinol, m-tolylmethanol, benzenemethanol, 3-methyl, unii-ttg38p37u6, m-methylbenzyl alcohol, 3-methylbenzylalcohol, m-methyl benzyl alcohol, 3-methyl-benzenemethanol
PubChem CID 11476
ChEBI CHEBI:27995
IUPAC Name (3-methylphenyl)methanol
SMILES CC1=CC(=CC=C1)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2087514
View Documents
Thermo Scientific Chemicals
B2087514
25 g
Each for $131.63
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

587-03-1
122.167
JJCKHVUTVOPLBV-UHFFFAOYSA-N
11476
(3-methylphenyl)methanol
C8H10O
MFCD00004646
3-methylbenzyl alcohol, 3-methylphenyl methanol, m-tolyl carbinol, m-tolylmethanol, benzenemethanol, 3-methyl, unii-ttg38p37u6, m-methylbenzyl alcohol, 3-methylbenzylalcohol, m-methyl benzyl alcohol, 3-methyl-benzenemethanol
CHEBI:27995
CC1=CC(=CC=C1)CO

Specifications

587-03-1
216°C to 217°C
C8H10O
MFCD00004646
2324516
JJCKHVUTVOPLBV-UHFFFAOYSA-N
(3-methylphenyl)methanol
11476
122.17
3-Methylbenzyl alcohol
1.013
105°C (221°F)
1.534
25 g
3-methylbenzyl alcohol, 3-methylphenyl methanol, m-tolyl carbinol, m-tolylmethanol, benzenemethanol, 3-methyl, unii-ttg38p37u6, m-methylbenzyl alcohol, 3-methylbenzylalcohol, m-methyl benzyl alcohol, 3-methyl-benzenemethanol
CC1=CC(=CC=C1)CO
122.167
CHEBI:27995
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-595-3

RTECSNumber : DA4937010

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only