missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
$67.11 - $471.60
Chemical Identifiers
| CAS | 591-23-1 |
|---|---|
| Molecular Formula | C7H14O |
| Molecular Weight (g/mol) | 114.19 |
| MDL Number | MFCD00001446 |
| InChI Key | HTSABYAWKQAHBT-UHFFFAOYNA-N |
| Synonym | 3-methylcyclohexanol, m-methylcyclohexanol, cyclohexanol, 3-methyl, hexahydro-m-cresol, cyclohexanol, m-methyl, 3-methylcyclohexanol, mixed isomers, 3-methyl-cyclohexanol, cis-3-methylcyclohexanol, trans-3-methylcyclohexanol, 3-methylcyclohexanol, mixture of cis and trans |
| PubChem CID | 11566 |
| IUPAC Name | 3-methylcyclohexan-1-ol |
| SMILES | CC1CCCC(O)C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M019525ML
|
TCI America
M019525ML |
25 mL |
Each for $67.11
|
|
|||||
|
M0195500ML
|
TCI America
M0195500ML |
500 mL |
Each for $471.60
|
|
|||||
Chemical Identifiers
| 591-23-1 | |
| 114.19 | |
| HTSABYAWKQAHBT-UHFFFAOYNA-N | |
| 11566 | |
| CC1CCCC(O)C1 |
| C7H14O | |
| MFCD00001446 | |
| 3-methylcyclohexanol, m-methylcyclohexanol, cyclohexanol, 3-methyl, hexahydro-m-cresol, cyclohexanol, m-methyl, 3-methylcyclohexanol, mixed isomers, 3-methyl-cyclohexanol, cis-3-methylcyclohexanol, trans-3-methylcyclohexanol, 3-methylcyclohexanol, mixture of cis and trans | |
| 3-methylcyclohexan-1-ol |
Specifications
| 591-23-1 | |
| C7H14O | |
| 25 mL | |
| HTSABYAWKQAHBT-UHFFFAOYNA-N | |
| 3-methylcyclohexan-1-ol | |
| 11566 | |
| ≥98.0% (GC) | |
| 3-Methylcyclohexanol (cis- and trans- mixture) |
| Colorless | |
| MFCD00001446 | |
| 3-methylcyclohexanol, m-methylcyclohexanol, cyclohexanol, 3-methyl, hexahydro-m-cresol, cyclohexanol, m-methyl, 3-methylcyclohexanol, mixed isomers, 3-methyl-cyclohexanol, cis-3-methylcyclohexanol, trans-3-methylcyclohexanol, 3-methylcyclohexanol, mixture of cis and trans | |
| CC1CCCC(O)C1 | |
| 114.19 | |
| 114.19 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2301
RTECSNumber : GW0200000
TSCA : Yes