Learn More
3-Methylcyclohexanone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 126631000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Methylcyclohexanone | |
591-24-2 | |
100.0 | |
Colorless to Yellow | |
169.0°C to 170.0°C | |
Authentic | |
Glass bottle | |
1.4440 to 1.4460 | |
MFCD00001639 | |
07, 15 | |
3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s | |
UJBOOUHRTQVGRU-UHFFFAOYNA-N | |
3-methylcyclohexan-1-one | |
11567 | |
97% |
97% | |
96.0 | |
-73.0°C | |
0.9150g/mL | |
48°C | |
97% | |
C7H12O | |
CH3C6H9(=O) | |
100 g | |
0.915 | |
Solubility in water: insoluble | |
CC1CCCC(=O)C1 | |
112.17 | |
112.17 | |
Liquid |
Chemical Identifiers
591-24-2 | |
112.17 | |
UJBOOUHRTQVGRU-UHFFFAOYNA-N | |
11567 | |
CC1CCCC(=O)C1 |
C7H12O | |
MFCD00001639 | |
3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s | |
3-methylcyclohexan-1-one |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to f
GHS Signal Word: Warning
EINECSNumber : 209-710-7
RTECSNumber : GW1750100
TSCA : TSCA
RUO – Research Use Only