missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylcyclohexanone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 126635000

 View more versions of this product

Catalog No. AC126635000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methylcyclohexanone
591-24-2
100.0
Colorless to Yellow
169.0°C to 170.0°C
Authentic
Glass bottle
1.4440 to 1.4460
MFCD00001639
07, 15
3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s
UJBOOUHRTQVGRU-UHFFFAOYNA-N
3-methylcyclohexan-1-one
11567
97%
97%
96.0
-73.0°C
0.9150g/mL
48°C
97%
C7H12O
CH3C6H9(=O)
500 g
0.915
Solubility in water: insoluble
CC1CCCC(=O)C1
112.17
112.17
Liquid

Chemical Identifiers

591-24-2
112.17
UJBOOUHRTQVGRU-UHFFFAOYNA-N
11567
CC1CCCC(=O)C1
C7H12O
MFCD00001639
3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s
3-methylcyclohexan-1-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to f

GHS Signal Word: Warning

EINECSNumber : 209-710-7

RTECSNumber : GW1750100

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only