missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methylcyclopentane-1,2-dione, 98+%, Thermo Scientific Chemicals
$56.45 - $302.18
Chemical Identifiers
CAS | 765-70-8 |
---|---|
Molecular Formula | C6H8O2 |
Molecular Weight (g/mol) | 112.128 |
MDL Number | MFCD00001417 |
InChI Key | OACYKCIZDVVNJL-UHFFFAOYSA-N |
Synonym | 3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone |
PubChem CID | 61209 |
IUPAC Name | 3-methylcyclopentane-1,2-dione |
SMILES | CC1CCC(=O)C1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1025509
|
Thermo Scientific Chemicals
A1025509 |
10 g |
N/A
|
|
|||||
AAA1025518
|
Thermo Scientific Chemicals
A1025518 |
50 g |
N/A
|
|
|||||
AAA1025522
|
Thermo Scientific Chemicals
A1025522 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
765-70-8 | |
112.128 | |
OACYKCIZDVVNJL-UHFFFAOYSA-N | |
61209 | |
CC1CCC(=O)C1=O |
C6H8O2 | |
MFCD00001417 | |
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone | |
3-methylcyclopentane-1,2-dione |
Specifications
765-70-8 | |
C6H8O2 | |
10 g | |
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone | |
CC1CCC(=O)C1=O | |
112.128 | |
112.13 | |
3-Methylcyclopentane-1,2-dione |
103°C to 107°C | |
MFCD00001417 | |
2076520 | |
OACYKCIZDVVNJL-UHFFFAOYSA-N | |
3-methylcyclopentane-1,2-dione | |
61209 | |
≥98% |
Safety and Handling
EINECSNumber : 212-154-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only