missing translation for 'onlineSavingsMsg'
Learn More

3-Methylglutaric anhydride, 97%

CAS: 4166-53-4 | C6H8O3 | 128.13 g/mol

$297.43 - $297.43

Chemical Identifiers

CAS 4166-53-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00006683
InChI Key MGICRVTUCPFQQZ-UHFFFAOYSA-N
Synonym 3-methylglutaric anhydride, 4-methyldihydro-2h-pyran-2,6 3h-dione, 3-methylglutaricanhydride, .beta.-methylglutaric anhydride, acmc-1ago6, 2h-pyran-2,6 3h-dione, dihydro-4-methyl, 3-methyl glutaric anhydride, beta-methylglutaric anhydride, 4-methyltetrahydropyran-2,6-dione
PubChem CID 77828
IUPAC Name 4-methyloxane-2,6-dione
SMILES CC1CC(=O)OC(=O)C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC126870250
SDS View Documents
Thermo Scientific Chemicals
126870250
25 g Glass bottle
Each for $297.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4166-53-4
128.13
MGICRVTUCPFQQZ-UHFFFAOYSA-N
77828
CC1CC(=O)OC(=O)C1
C6H8O3
MFCD00006683
3-methylglutaric anhydride, 4-methyldihydro-2h-pyran-2,6 3h-dione, 3-methylglutaricanhydride, .beta.-methylglutaric anhydride, acmc-1ago6, 2h-pyran-2,6 3h-dione, dihydro-4-methyl, 3-methyl glutaric anhydride, beta-methylglutaric anhydride, 4-methyltetrahydropyran-2,6-dione
4-methyloxane-2,6-dione

Specifications

4166-53-4
100.0
White
>110°C
97%
C6H8O3
25 g
3-methylglutaric anhydride, 4-methyldihydro-2h-pyran-2,6 3h-dione, 3-methylglutaricanhydride, .beta.-methylglutaric anhydride, acmc-1ago6, 2h-pyran-2,6 3h-dione, dihydro-4-methyl, 3-methyl glutaric anhydride, beta-methylglutaric anhydride, 4-methyltetrahydropyran-2,6-dione
MGICRVTUCPFQQZ-UHFFFAOYSA-N
4-methyloxane-2,6-dione
77828
97%
3-Methylglutaric anhydride, 97%
96.0
40°C to 45°C
180°C to 182°C (25.0 mmHg)
Authentic
Glass bottle
MFCD00006683
17, 415
Solubility in water: may decompose
CC1CC(=O)OC(=O)C1
128.13
128.13
Crystalline Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-020-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only