missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methylhexane 95.0+%, TCI America™
Supplier: TCI America M034025ML
Specifications
3-Methylhexane | |
Colorless | |
C7H16 | |
25 mL | |
hexane, 3-methyl, 2-ethylpentane, 3-methyl hexane, 3-methyl-hexane, 3-methylhexane, dsstox_cid_8974, acmc-209m8a, s-+-3-methylhexane, dsstox_rid_80156, dsstox_gsid_44334 | |
CCCC(C)CC | |
100.205 | |
100.21 | |
Liquid |
589-34-4 | |
91°C | |
MFCD00009408 | |
1206 | |
VLJXXKKOSFGPHI-UHFFFAOYSA-N | |
3-methylhexane | |
11507 | |
≥95.0% (GC) |
Chemical Identifiers
589-34-4 | |
100.205 | |
VLJXXKKOSFGPHI-UHFFFAOYSA-N | |
11507 | |
CCCC(C)CC |
C7H16 | |
MFCD00009408 | |
hexane, 3-methyl, 2-ethylpentane, 3-methyl hexane, 3-methyl-hexane, 3-methylhexane, dsstox_cid_8974, acmc-209m8a, s-+-3-methylhexane, dsstox_rid_80156, dsstox_gsid_44334 | |
3-methylhexane |
Safety and Handling
EINECSNumber : (2)-0007
TSCA : Yes