Learn More
3-Methylpyridine-2-carboxaldehyde, 96%, Thermo Scientific Chemicals
$185.88 - $640.78
Chemical Identifiers
CAS | 55589-47-4 |
---|---|
Molecular Formula | C7H7NO |
Molecular Weight (g/mol) | 121.139 |
MDL Number | MFCD02181144 |
InChI Key | JDYVLWWFVYNMTN-UHFFFAOYSA-N |
Synonym | 3-methylpicolinaldehyde, 3-methyl-2-pyridinecarboxaldehyde, 3-methylpyridine-2-carboxaldehyde, 3-methyl-2-pyridinaldehyde, 2-formyl-3-picoline, 3-methyl-pyridine-2-carbaldehyde, 2-formyl-3-methylpyridine, pyridinecarboxaldehyde, methyl, 2-pyridinecarboxaldehyde, 3-methyl, 3-methyl-2-pyridineformaldehyde |
PubChem CID | 1501874 |
IUPAC Name | 3-methylpyridine-2-carbaldehyde |
SMILES | CC1=C(N=CC=C1)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5616803
|
Thermo Scientific Chemicals
H5616803 |
1 g |
Each for $185.88
|
|
|||||
AAH5616806
|
Thermo Scientific Chemicals
H5616806 |
5 g |
Each for $640.78
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents. Store at room temperature.
Chemical Identifiers
55589-47-4 | |
121.139 | |
JDYVLWWFVYNMTN-UHFFFAOYSA-N | |
1501874 | |
CC1=C(N=CC=C1)C=O |
C7H7NO | |
MFCD02181144 | |
3-methylpicolinaldehyde, 3-methyl-2-pyridinecarboxaldehyde, 3-methylpyridine-2-carboxaldehyde, 3-methyl-2-pyridinaldehyde, 2-formyl-3-picoline, 3-methyl-pyridine-2-carbaldehyde, 2-formyl-3-methylpyridine, pyridinecarboxaldehyde, methyl, 2-pyridinecarboxaldehyde, 3-methyl, 3-methyl-2-pyridineformaldehyde | |
3-methylpyridine-2-carbaldehyde |
Specifications
55589-47-4 | |
73°C (163°F) | |
1.538 | |
1 g | |
Slightly soluble in water. | |
CC1=C(N=CC=C1)C=O | |
121.139 | |
121.14 | |
3-Methylpyridine-2-carboxaldehyde |
1.08 | |
C7H7NO | |
MFCD02181144 | |
3-methylpicolinaldehyde, 3-methyl-2-pyridinecarboxaldehyde, 3-methylpyridine-2-carboxaldehyde, 3-methyl-2-pyridinaldehyde, 2-formyl-3-picoline, 3-methyl-pyridine-2-carbaldehyde, 2-formyl-3-methylpyridine, pyridinecarboxaldehyde, methyl, 2-pyridinecarboxaldehyde, 3-methyl, 3-methyl-2-pyridineformaldehyde | |
JDYVLWWFVYNMTN-UHFFFAOYSA-N | |
3-methylpyridine-2-carbaldehyde | |
1501874 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only