Learn More
3-Methylstyrene, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 100-80-1 |
---|---|
Molecular Formula | C9H10 |
Molecular Weight (g/mol) | 118.18 |
InChI Key | JZHGRUMIRATHIU-UHFFFAOYSA-N |
Synonym | 3-methylstyrene, 3-vinyltoluene, 1-methyl-3-vinylbenzene, m-vinyltoluene, m-methylstyrene, benzene, 1-ethenyl-3-methyl, styrene, m-methyl, m-methyl styrene, meta-methylstyrene, unii-ffk4i9430g |
PubChem CID | 7529 |
IUPAC Name | 1-ethenyl-3-methylbenzene |
SMILES | CC1=CC=CC(=C1)C=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC450870010
|
Thermo Scientific Chemicals
450870010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC450870050
|
Thermo Scientific Chemicals
450870050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
100-80-1 | |
118.18 | |
3-methylstyrene, 3-vinyltoluene, 1-methyl-3-vinylbenzene, m-vinyltoluene, m-methylstyrene, benzene, 1-ethenyl-3-methyl, styrene, m-methyl, m-methyl styrene, meta-methylstyrene, unii-ffk4i9430g | |
1-ethenyl-3-methylbenzene |
C9H10 | |
JZHGRUMIRATHIU-UHFFFAOYSA-N | |
7529 | |
CC1=CC=CC(=C1)C=C |
Specifications
100-80-1 | |
51°C | |
94% min. (GC) | |
C9H10 | |
3-methylstyrene, 3-vinyltoluene, 1-methyl-3-vinylbenzene, m-vinyltoluene, m-methylstyrene, benzene, 1-ethenyl-3-methyl, styrene, m-methyl, m-methyl styrene, meta-methylstyrene, unii-ffk4i9430g | |
CC1=CC=CC(=C1)C=C | |
118.18 | |
118.18 | |
3-Methylstyrene |
170.0°C to 171.0°C | |
Authentic | |
Glass Bottle | |
1g | |
JZHGRUMIRATHIU-UHFFFAOYSA-N | |
1-ethenyl-3-methylbenzene | |
7529 | |
95% |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 202-889-2
RUO â Research Use Only