Learn More
3-(Methylthio)benzoic acid, 97%, Thermo Scientific Chemicals
$90.03 - $933.18
Chemical Identifiers
CAS | 825-99-0 |
---|---|
Molecular Formula | C8H8O2S |
Molecular Weight (g/mol) | 168.21 |
MDL Number | MFCD00156993 |
InChI Key | PZGADOOBMVLBJE-UHFFFAOYSA-N |
Synonym | 3-methylthio benzoic acid, 3-methylsulfanyl benzoic acid, benzoic acid, 3-methylthio, benzoic acid, m-methylthio, 3-mercaptobenzoic acid s-methyl ether, 3-methylsulfanyl-benzoic acid, 3-carboxythioanisole, pubchem10679 |
PubChem CID | 220329 |
IUPAC Name | 3-methylsulfanylbenzoic acid |
SMILES | CSC1=CC=CC(=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2720003
|
Thermo Scientific Chemicals
H2720003 |
1 g |
Each for $90.03
|
|
|||||
AAH2720006
|
Thermo Scientific Chemicals
H2720006 |
5 g |
Each for $294.01
|
|
|||||
AAH2720014
|
Thermo Scientific Chemicals
H2720014 |
25 g |
Each for $933.18
|
|
|||||
Description
It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
825-99-0 | |
168.21 | |
PZGADOOBMVLBJE-UHFFFAOYSA-N | |
220329 | |
CSC1=CC=CC(=C1)C(=O)O |
C8H8O2S | |
MFCD00156993 | |
3-methylthio benzoic acid, 3-methylsulfanyl benzoic acid, benzoic acid, 3-methylthio, benzoic acid, m-methylthio, 3-mercaptobenzoic acid s-methyl ether, 3-methylsulfanyl-benzoic acid, 3-carboxythioanisole, pubchem10679 | |
3-methylsulfanylbenzoic acid |
Specifications
825-99-0 | |
C8H8O2S | |
1 g | |
PZGADOOBMVLBJE-UHFFFAOYSA-N | |
3-methylsulfanylbenzoic acid | |
220329 | |
97% |
126°C to 132°C | |
MFCD00156993 | |
3-methylthio benzoic acid, 3-methylsulfanyl benzoic acid, benzoic acid, 3-methylthio, benzoic acid, m-methylthio, 3-mercaptobenzoic acid s-methyl ether, 3-methylsulfanyl-benzoic acid, 3-carboxythioanisole, pubchem10679 | |
CSC1=CC=CC(=C1)C(=O)O | |
168.21 | |
168.21 | |
3-(Methylthio)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only