missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methylthio)phenyl isocyanate, 96%, Thermo Scientific Chemicals

$86.92 - $273.93

Chemical Identifiers

CAS 28479-19-8
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00013863
InChI Key BKJABLMNBSVKCV-UHFFFAOYSA-N
Synonym 3-methylthio phenyl isocyanate, 1-isocyanato-3-methylthio benzene, 3-methylthiophenyl isocyanate, 1-isocyanato-3-methylsulfanyl benzene, 1-isocyanato-3-methylsulfanyl-benzene, 3-methylthio phenylisocyanate, benzene, 1-isocyanato-3-methylthio, 3-methylthiobenzenisocyanate, acmc-20anig, pubchem14329
PubChem CID 141418
IUPAC Name 1-isocyanato-3-methylsulfanylbenzene
SMILES CSC1=CC=CC(=C1)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1200803
View Documents
Thermo Scientific Chemicals
L1200803
1 g
Each for $86.92
 
AAL1200806
View Documents
Thermo Scientific Chemicals
L1200806
5 g
Each for $273.93
 
Description

Description

3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines and alcohols. It is sensitive to moisture. Store under dry inert gas.
Specifications

Chemical Identifiers

28479-19-8
165.21
BKJABLMNBSVKCV-UHFFFAOYSA-N
141418
CSC1=CC=CC(=C1)N=C=O
C8H7NOS
MFCD00013863
3-methylthio phenyl isocyanate, 1-isocyanato-3-methylthio benzene, 3-methylthiophenyl isocyanate, 1-isocyanato-3-methylsulfanyl benzene, 1-isocyanato-3-methylsulfanyl-benzene, 3-methylthio phenylisocyanate, benzene, 1-isocyanato-3-methylthio, 3-methylthiobenzenisocyanate, acmc-20anig, pubchem14329
1-isocyanato-3-methylsulfanylbenzene

Specifications

28479-19-8
129°C to 130°C (5 mmHg)
C8H7NOS
MFCD00013863
UN2206
Moisture sensitive
Hydrolyzes in water.
CSC1=CC=CC(=C1)N=C=O
165.21
165.22
3-(Methylthio)phenyl isocyanate
1.199
113°C (235°F)
1.603
1 g
387507
3-methylthio phenyl isocyanate, 1-isocyanato-3-methylthio benzene, 3-methylthiophenyl isocyanate, 1-isocyanato-3-methylsulfanyl benzene, 1-isocyanato-3-methylsulfanyl-benzene, 3-methylthio phenylisocyanate, benzene, 1-isocyanato-3-methylthio, 3-methylthiobenzenisocyanate, acmc-20anig, pubchem14329
BKJABLMNBSVKCV-UHFFFAOYSA-N
1-isocyanato-3-methylsulfanylbenzene
141418
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c

H302-H317-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only