missing translation for 'onlineSavingsMsg'
Learn More

3-methylthiophene-2-carbonyl Chloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 61341-26-2
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00130091
InChI Key AJSVPEVDFBYRCH-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
PubChem CID 2737605
IUPAC Name 3-methylthiophene-2-carbonyl chloride
SMILES CC1=C(SC=C1)C(Cl)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC380400020
SDS View Documents
Thermo Scientific Chemicals
380400020
2g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC380400100
SDS View Documents
Thermo Scientific Chemicals
380400100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

61341-26-2
160.62
AJSVPEVDFBYRCH-UHFFFAOYSA-N
2737605
CC1=C(SC=C1)C(Cl)=O
C6H5ClOS
MFCD00130091
3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
3-methylthiophene-2-carbonyl chloride

Specifications

61341-26-2
100.0
>110°C
98%
C6H5ClOS
MFCD00130091
3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
CC1=C(SC=C1)C(Cl)=O
160.62
160.62
3-Methylthiophene-2-carbonyl chloride
97.5
216°C to 220°C
Authentic
Glass bottle
1.5830 to 1.585
2g
AJSVPEVDFBYRCH-UHFFFAOYSA-N
3-methylthiophene-2-carbonyl chloride
2737605
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only