missing translation for 'onlineSavingsMsg'
Learn More

3-(N-BOC-aminomethyl)-5-bromopyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 943722-24-5
Molecular Formula C11H15BrN2O2
Molecular Weight (g/mol) 287.16
InChI Key VZPOHASZICQBDX-UHFFFAOYSA-N
Synonym 3-n-boc-aminomethyl-5-bromopyridine, tert-butyl 5-bromopyridin-3-yl methylcarbamate, tert-butyl 5-bromopyridin-3-yl methyl carbamate, tert-butyl n-5-bromopyridin-3-yl methyl carbamate, 3-bromo-5-tert-butoxycarbonyl amino methyl pyridine, 3-boc-aminomethyl-5-bromopyridine, 5-bromo-3-boc-amino methyl pyridine, 5-bromo-3-n-tert-butyl carbamatomethylpyridine, 3-aminomethyl-5-bromopyridine, 3-boc protected
PubChem CID 46739290
IUPAC Name tert-butyl N-[(5-bromopyridin-3-yl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC(=CN=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444060010
SDS View Documents
Thermo Scientific Chemicals
444060010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444060050
SDS View Documents
Thermo Scientific Chemicals
444060050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

943722-24-5
287.16
3-n-boc-aminomethyl-5-bromopyridine, tert-butyl 5-bromopyridin-3-yl methylcarbamate, tert-butyl 5-bromopyridin-3-yl methyl carbamate, tert-butyl n-5-bromopyridin-3-yl methyl carbamate, 3-bromo-5-tert-butoxycarbonyl amino methyl pyridine, 3-boc-aminomethyl-5-bromopyridine, 5-bromo-3-boc-amino methyl pyridine, 5-bromo-3-n-tert-butyl carbamatomethylpyridine, 3-aminomethyl-5-bromopyridine, 3-boc protected
tert-butyl N-[(5-bromopyridin-3-yl)methyl]carbamate
C11H15BrN2O2
VZPOHASZICQBDX-UHFFFAOYSA-N
46739290
CC(C)(C)OC(=O)NCC1=CC(=CN=C1)Br

Specifications

943722-24-5
Glass Bottle
1g
VZPOHASZICQBDX-UHFFFAOYSA-N
tert-butyl N-[(5-bromopyridin-3-yl)methyl]carbamate
46739290
97%
96% min. (GC)
C11H15BrN2O2
3-n-boc-aminomethyl-5-bromopyridine, tert-butyl 5-bromopyridin-3-yl methylcarbamate, tert-butyl 5-bromopyridin-3-yl methyl carbamate, tert-butyl n-5-bromopyridin-3-yl methyl carbamate, 3-bromo-5-tert-butoxycarbonyl amino methyl pyridine, 3-boc-aminomethyl-5-bromopyridine, 5-bromo-3-boc-amino methyl pyridine, 5-bromo-3-n-tert-butyl carbamatomethylpyridine, 3-aminomethyl-5-bromopyridine, 3-boc protected
CC(C)(C)OC(=O)NCC1=CC(=CN=C1)Br
287.16
287.16
3-(N-BOC-aminomethyl)-5-bromopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only