missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[3-(N,N-Dimethylamino)propyl]trimethoxysilane 96.0+%, TCI America™
$97.89 - $315.21
Chemical Identifiers
CAS | 2530-86-1 |
---|---|
Molecular Formula | C8H21NO3Si |
Molecular Weight (g/mol) | 207.35 |
MDL Number | MFCD00053674 |
InChI Key | QIOYHIUHPGORLS-UHFFFAOYSA-N |
Synonym | N,N-Dimethyl-3-(trimethoxysilyl)propylamine |
PubChem CID | 75668 |
IUPAC Name | dimethyl[3-(trimethoxysilyl)propyl]amine |
SMILES | CO[Si](CCCN(C)C)(OC)OC |
Chemical Identifiers
2530-86-1 | |
207.35 | |
QIOYHIUHPGORLS-UHFFFAOYSA-N | |
75668 | |
CO[Si](CCCN(C)C)(OC)OC |
C8H21NO3Si | |
MFCD00053674 | |
N,N-Dimethyl-3-(trimethoxysilyl)propylamine | |
dimethyl[3-(trimethoxysilyl)propyl]amine |
Specifications
2530-86-1 | |
42°C | |
MFCD00053674 | |
1760 | |
QIOYHIUHPGORLS-UHFFFAOYSA-N | |
dimethyl[3-(trimethoxysilyl)propyl]amine | |
75668 | |
≥96.0% (GC,T) | |
[3-(N,N-Dimethylamino)propyl]trimethoxysilane |
Yellow | |
C8H21NO3Si | |
5 mL | |
N,N-Dimethyl-3-(trimethoxysilyl)propylamine | |
CO[Si](CCCN(C)C)(OC)OC | |
207.35 | |
207.35 | |
Liquid |
Safety and Handling
TSCA : Yes