Learn More
3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 427740250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Phase transfer catalystSpecifications
3-(N, N-Dimethylmyristylammonio)propanesulfonate | |
96.0 | |
60.57 to 64.96 % C 10.97 to 11.76 % H 3.72 to 3.99 % N | |
97% | |
C19H41NO3S | |
25g | |
BHATUINFZWUDIX-UHFFFAOYSA-N | |
3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate | |
84705 | |
363.61 |
14933-09-6 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00036910 | |
myristyl sulfobetaine, zwittergent 3-14, n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dimethyl tetradecyl ammonio propane-1-sulfonate, 3-n,n-dimethylmyristylammonio propanesulfonate, dimethyl 3-sulphonatopropyl tetradecylammonium, zwittergent™ 3-14 detergent, 3-myristyldimethylammonio propanesulfonate, 3-n,n-dimethyltetradecylammonio propanesulfonate | |
CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] | |
363.61 | |
CHEBI:10129 | |
97% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
EINECSNumber : 239-003-9
RUO â Research Use Only