missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitro-L-phenylalanine, 95%
CAS: 19883-74-0 | C9H10N2O4 | 210.19 g/mol
$94.97 - $1064.48
Chemical Identifiers
| CAS | 19883-74-0 |
|---|---|
| Molecular Formula | C9H10N2O4 |
| Molecular Weight (g/mol) | 210.19 |
| MDL Number | MFCD01860661 |
| InChI Key | YTHDRUZHNYKZGF-UHFFFAOYNA-N |
| Synonym | 3-nitro-l-phenylalanine, s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophenylalanine, 3-nitrophenylalanine, l-3-nitro phenylalanine, 2s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophe, melphalan impurity 2, l-3-no2-phe-oh, ncistruc1_000602 |
| PubChem CID | 2761812 |
| IUPAC Name | 2-amino-3-(3-nitrophenyl)propanoic acid |
| SMILES | NC(CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH63029MD
|
Thermo Scientific Chemicals
H63029MD |
250 mg |
Each for $94.97
|
|
|||||
|
AAH6302903
|
Thermo Scientific Chemicals
H6302903 |
1 g |
Each for $248.90
|
|
|||||
|
AAH6302906
|
Thermo Scientific Chemicals
H6302906 |
5 g |
Each for $1,064.48
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19883-74-0 | |
| 210.19 | |
| YTHDRUZHNYKZGF-UHFFFAOYNA-N | |
| 2761812 | |
| NC(CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O |
| C9H10N2O4 | |
| MFCD01860661 | |
| 3-nitro-l-phenylalanine, s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophenylalanine, 3-nitrophenylalanine, l-3-nitro phenylalanine, 2s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophe, melphalan impurity 2, l-3-no2-phe-oh, ncistruc1_000602 | |
| 2-amino-3-(3-nitrophenyl)propanoic acid |
Specifications
| 19883-74-0 | |
| C9H10N2O4 | |
| 3-nitro-l-phenylalanine, s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophenylalanine, 3-nitrophenylalanine, l-3-nitro phenylalanine, 2s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophe, melphalan impurity 2, l-3-no2-phe-oh, ncistruc1_000602 | |
| NC(CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O | |
| 250 mg | |
| 2761812 | |
| 95% | |
| 3-Nitro-L-phenylalanine |
| White | |
| MFCD01860661 | |
| YTHDRUZHNYKZGF-UHFFFAOYNA-N | |
| 2-amino-3-(3-nitrophenyl)propanoic acid | |
| 210.19 | |
| 210.19 | |
| Powder |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only