missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitroaniline, 98%, Thermo Scientific Chemicals

$112.00 - $190.47

Chemical Identifiers

CAS 99-09-2
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007782
InChI Key XJCVRTZCHMZPBD-UHFFFAOYSA-N
Synonym m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine
PubChem CID 7423
IUPAC Name 3-nitroaniline
SMILES NC1=CC=CC(=C1)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128361000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
128361000
100 g Glass bottle
Each for $112.00
 
AC128365000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
128365000
500 g Glass bottle
Each for $190.47
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

99-09-2
138.13
XJCVRTZCHMZPBD-UHFFFAOYSA-N
7423
NC1=CC=CC(=C1)[N+]([O-])=O
C6H6N2O2
MFCD00007782
m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine
3-nitroaniline

Specifications

99-09-2
100.0
Orange to Yellow
196°C
97.5% min. (GC)
C6H6N2O2
MFCD00007782
12, 698
m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine
XJCVRTZCHMZPBD-UHFFFAOYSA-N
3-nitroaniline
7423
98%
3-Nitroaniline, 99+%
97.5
111.0°C to 114.0°C
306.0°C
Authentic
Glass bottle
O2NC6H4NH2
100 g
15, 6668
Solubility in water: 1.25g/L in water. Other solubilities: soluble in alcohol,ether,methanol and ethanol,1 g soluble in 20 mL alcohol,1 g soluble in 18 mL ether,1 g soluble in 11.5 mL methanol
NC1=CC=CC(=C1)[N+]([O-])=O
138.13
138.13
Crystalline Powder, Crystals or Chunks
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove

GHS Signal Word: Danger

EINECSNumber : 202-729-1

RTECSNumber : BY6825000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only