missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Nitrobenzeneboronic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1333614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Nitrobenzeneboronic acid | |
∼285°C (decomposition) | |
MFCD00007193 | |
2938638 | |
ZNRGSYUVFVNSAW-UHFFFAOYSA-N | |
(3-nitrophenyl)boronic acid | |
1677 | |
98% |
13331-27-6 | |
C6H6BNO4 | |
25 g | |
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol | |
OB(O)C1=CC=CC(=C1)[N+]([O-])=O | |
166.93 | |
166.93 |
Chemical Identifiers
13331-27-6 | |
166.93 | |
ZNRGSYUVFVNSAW-UHFFFAOYSA-N | |
1677 | |
OB(O)C1=CC=CC(=C1)[N+]([O-])=O |
C6H6BNO4 | |
MFCD00007193 | |
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol | |
(3-nitrophenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : CY8980000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only