missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrobenzyl alcohol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1504022

 View more versions of this product

Catalog No. AAA1504022


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Nitrobenzyl alcohol
29°C to 33°C
175°C to 180°C (3 mmHg)
C7H7NO3
MFCD00007273
2044769
CWNPOQFCIIFQDM-UHFFFAOYSA-N
(3-nitrophenyl)methanol
69267
98%
619-25-0
1.3
>110°C (230°F)
1.5755
100 g
3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol
C1=CC(=CC(=C1)[N+](=O)[O-])CO
153.137
153.14

Chemical Identifiers

619-25-0
153.137
CWNPOQFCIIFQDM-UHFFFAOYSA-N
69267
C1=CC(=CC(=C1)[N+](=O)[O-])CO
C7H7NO3
MFCD00007273
3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol
(3-nitrophenyl)methanol
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-588-2

RTECSNumber : DP0657000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only