missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrocinnamic Acid 98.0+%, TCI America™

$39.14 - $100.67

Chemical Identifiers

CAS 555-68-0
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00007277
InChI Key WWXMVRYHLZMQIG-SNAWJCMRSA-N
Synonym 3-nitrocinnamic acid, m-nitrocinnamic acid, 3-nitro-cinnamic acid, cinnamic acid, m-nitro, trans-3-nitrocinnamic acid, 3-nitrobenzenepropenoic acid, e-3-3-nitrophenyl-2-propenoic acid, 2e-3-3-nitrophenyl prop-2-enoic acid, 2e-3-3-nitrophenyl acrylic acid, unii-z5wu65j930
PubChem CID 687126
ChEBI CHEBI:60706
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C=CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N03545G
View Documents
TCI America
N03545G
5 g
Each for $39.14
Only null left
 
N035425G
View Documents
TCI America
N035425G
25 g
Each for $100.67
Only null left
 
Description

Description

Specifications

Chemical Identifiers

555-68-0
193.158
WWXMVRYHLZMQIG-SNAWJCMRSA-N
687126
C1=CC(=CC(=C1)[N+](=O)[O-])C=CC(=O)O
C9H7NO4
MFCD00007277
3-nitrocinnamic acid, m-nitrocinnamic acid, 3-nitro-cinnamic acid, cinnamic acid, m-nitro, trans-3-nitrocinnamic acid, 3-nitrobenzenepropenoic acid, e-3-3-nitrophenyl-2-propenoic acid, 2e-3-3-nitrophenyl prop-2-enoic acid, 2e-3-3-nitrophenyl acrylic acid, unii-z5wu65j930
CHEBI:60706

Specifications

555-68-0
White-Yellow
MFCD00007277
3-nitrocinnamic acid, m-nitrocinnamic acid, 3-nitro-cinnamic acid, cinnamic acid, m-nitro, trans-3-nitrocinnamic acid, 3-nitrobenzenepropenoic acid, e-3-3-nitrophenyl-2-propenoic acid, 2e-3-3-nitrophenyl prop-2-enoic acid, 2e-3-3-nitrophenyl acrylic acid, unii-z5wu65j930
C1=CC(=CC(=C1)[N+](=O)[O-])C=CC(=O)O
687126
193.16
Crystalline Powder
205°C
C9H7NO4
5 g
WWXMVRYHLZMQIG-SNAWJCMRSA-N
193.158
CHEBI:60706
≥98.0% (T)
3-Nitrocinnamic Acid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : GE0352000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'