missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophthalimide, 98%, Thermo Scientific Chemicals

$71.96 - $681.16

Chemical Identifiers

CAS 603-62-3
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00041852
InChI Key BONIIQYTWOPUQI-UHFFFAOYSA-N
Synonym 3-nitrophthalimide, 4-nitroisoindoline-1,3-dione, phthalimide, 3-nitro, 1h-isoindole-1,3 2h-dione, 4-nitro, nitrophthalimide, 4-nitro-2,3-dihydro-1h-isoindole-1,3-dione, 4-nitro-1h-isoindole-1,3 2h-dione, 4-nitro-2h-benzo c azoline-1,3-dione, 3-nitro-phthalimide, pubchem12007
PubChem CID 11779
IUPAC Name 4-nitroisoindole-1,3-dione
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])C(=O)NC2=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1529106
View Documents
Thermo Scientific Chemicals
A1529106
5 g
Each for $71.96
 
AAA1529114
View Documents
Thermo Scientific Chemicals
A1529114
25 g
Each for $241.05
 
AAA1529122
View Documents
Thermo Scientific Chemicals
A1529122
100 g
Each for $681.16
 
Description

Description

3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents and bases.
Specifications

Chemical Identifiers

603-62-3
192.13
BONIIQYTWOPUQI-UHFFFAOYSA-N
11779
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=O)NC2=O
C8H4N2O4
MFCD00041852
3-nitrophthalimide, 4-nitroisoindoline-1,3-dione, phthalimide, 3-nitro, 1h-isoindole-1,3 2h-dione, 4-nitro, nitrophthalimide, 4-nitro-2,3-dihydro-1h-isoindole-1,3-dione, 4-nitro-1h-isoindole-1,3 2h-dione, 4-nitro-2h-benzo c azoline-1,3-dione, 3-nitro-phthalimide, pubchem12007
4-nitroisoindole-1,3-dione

Specifications

603-62-3
C8H4N2O4
5 g
3-nitrophthalimide, 4-nitroisoindoline-1,3-dione, phthalimide, 3-nitro, 1h-isoindole-1,3 2h-dione, 4-nitro, nitrophthalimide, 4-nitro-2,3-dihydro-1h-isoindole-1,3-dione, 4-nitro-1h-isoindole-1,3 2h-dione, 4-nitro-2h-benzo c azoline-1,3-dione, 3-nitro-phthalimide, pubchem12007
BONIIQYTWOPUQI-UHFFFAOYSA-N
4-nitroisoindole-1,3-dione
11779
98%
217°C to 221°C
MFCD00041852
179965
Insoluble in water.
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=O)NC2=O
192.13
192.13
3-Nitrophthalimide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-051-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only