missing translation for 'onlineSavingsMsg'
Learn More

3-Octen-2-one, 97%

CAS: 1669-44-9 | C8H14O | 126.199 g/mol

$85.78 - $1136.86

Chemical Identifiers

CAS 1669-44-9
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00015565
InChI Key ZCFOBLITZWHNNC-VOTSOKGWSA-N
Synonym 3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone
PubChem CID 5363229
IUPAC Name (E)-oct-3-en-2-one
SMILES CCCCC=CC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1465209
View Documents
Thermo Scientific Chemicals
A1465209
10 g
Each for $85.78
Only null left
 
AAA1465218
View Documents
Thermo Scientific Chemicals
A1465218
50 g
Each for $293.04
Only null left
 
AAA1465230
View Documents
Thermo Scientific Chemicals
A1465230
250 g
Each for $1,136.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1669-44-9
126.199
ZCFOBLITZWHNNC-VOTSOKGWSA-N
5363229
CCCCC=CC(=O)C
C8H14O
MFCD00015565
3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone
(E)-oct-3-en-2-one

Specifications

1669-44-9
60°C (16 mmHg)
Pleasant
1.448
10 g
1742230
ZCFOBLITZWHNNC-VOTSOKGWSA-N
(E)-oct-3-en-2-one
5363229
97%
0.85
54°C (129°F)
C8H14O
MFCD00015565
UN1224
3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone
CCCCC=CC(=O)C
126.199
126.2
3-Octen-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 216-793-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only