missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H62168MD

 View more versions of this product

Catalog No. AAH62168MD


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
C14H18BNO4
250 mg
ZXOSNHPLTJAXSA-UHFFFAOYSA-N
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
46856474
Solid
943994-02-3
MFCD12755796
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 3-oxo-2h,4h-benzo b 1,4 oxazine-6-boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,4-dihydro-1,4-benzoxazin-3-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4h-1,4-benzoxazin-3-one
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)OCC(=O)N3
275.111
275.11

Chemical Identifiers

943994-02-3
275.111
ZXOSNHPLTJAXSA-UHFFFAOYSA-N
46856474
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)OCC(=O)N3
C14H18BNO4
MFCD12755796
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 3-oxo-2h,4h-benzo b 1,4 oxazine-6-boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,4-dihydro-1,4-benzoxazin-3-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4h-1,4-benzoxazin-3-one
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions