missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 796061-08-0 |
---|---|
Molecular Formula | C19H24BNO4S |
Molecular Weight (g/mol) | 373.274 |
MDL Number | MFCD05663863 |
InChI Key | RGAZOXRZFCGICF-UHFFFAOYSA-N |
Synonym | 4-methyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl benzenesulfonamide, 3-toluene-4-sulfonylamino phenylboronic acid, pinacol ester, 3-p-toluenesulfonylamino phenylboronic acid pinacol ester, 3-p-toluenesulfonylamino benzeneboronic acid pinacol ester, 3-toluene-4-sulfonylamino phenylboronic acid pinacol ester, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-p-tolueneulfonamide, 3-toluene-4-sulfonylamino phenylboronic acid,pinacol ester, 3-toluene-4-sulfonylamino phenyl boronic acid, pinacol ester |
PubChem CID | 16217721 |
IUPAC Name | 4-methyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NS(=O)(=O)C3=CC=C(C=C3)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2723603
|
Thermo Scientific Chemicals
H2723603 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
796061-08-0 | |
373.274 | |
RGAZOXRZFCGICF-UHFFFAOYSA-N | |
16217721 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NS(=O)(=O)C3=CC=C(C=C3)C |
C19H24BNO4S | |
MFCD05663863 | |
4-methyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl benzenesulfonamide, 3-toluene-4-sulfonylamino phenylboronic acid, pinacol ester, 3-p-toluenesulfonylamino phenylboronic acid pinacol ester, 3-p-toluenesulfonylamino benzeneboronic acid pinacol ester, 3-toluene-4-sulfonylamino phenylboronic acid pinacol ester, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-p-tolueneulfonamide, 3-toluene-4-sulfonylamino phenylboronic acid,pinacol ester, 3-toluene-4-sulfonylamino phenyl boronic acid, pinacol ester | |
4-methyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide |
Specifications
796061-08-0 | |
C19H24BNO4S | |
1 g | |
RGAZOXRZFCGICF-UHFFFAOYSA-N | |
4-methyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide | |
16217721 | |
97% |
164°C to 168°C | |
MFCD05663863 | |
4-methyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl benzenesulfonamide, 3-toluene-4-sulfonylamino phenylboronic acid, pinacol ester, 3-p-toluenesulfonylamino phenylboronic acid pinacol ester, 3-p-toluenesulfonylamino benzeneboronic acid pinacol ester, 3-toluene-4-sulfonylamino phenylboronic acid pinacol ester, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-p-tolueneulfonamide, 3-toluene-4-sulfonylamino phenylboronic acid,pinacol ester, 3-toluene-4-sulfonylamino phenyl boronic acid, pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NS(=O)(=O)C3=CC=C(C=C3)C | |
373.274 | |
373.27 | |
3-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures