missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Pentadecylphenol 90.0+%, TCI America™
Supplier: TCI America P120225G
Specifications
3-Pentadecylphenol | |
50°C | |
C21H36O | |
25 g | |
PTFIPECGHSYQNR-UHFFFAOYSA-N | |
3-pentadecylphenol | |
68146 | |
≥90.0% (GC) |
501-24-6 | |
232°C | |
MFCD00002310 | |
3-n-pentadecylphenol, hydroginkgol, phenol, 3-pentadecyl, hydrocardanol, cyclogallipharaol, m-pentadecylphenol, tetrahydroanacardol, 3-pentadecyl-phenol, anacardol, tetrahydro, 3-pentadecyl phenol | |
CCCCCCCCCCCCCCCC1=CC=CC(O)=C1 | |
304.52 | |
304.52 | |
Crystalline Powder |
Chemical Identifiers
501-24-6 | |
304.52 | |
PTFIPECGHSYQNR-UHFFFAOYSA-N | |
68146 | |
CCCCCCCCCCCCCCCC1=CC=CC(O)=C1 |
C21H36O | |
MFCD00002310 | |
3-n-pentadecylphenol, hydroginkgol, phenol, 3-pentadecyl, hydrocardanol, cyclogallipharaol, m-pentadecylphenol, tetrahydroanacardol, 3-pentadecyl-phenol, anacardol, tetrahydro, 3-pentadecyl phenol | |
3-pentadecylphenol |
Safety and Handling
EINECSNumber : (3)-0511
TSCA : Yes