missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pentanol, 98+%, Thermo Scientific Chemicals

$130.03 - $540.55

Chemical Identifiers

CAS 584-02-1
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004574
InChI Key AQIXEPGDORPWBJ-UHFFFAOYSA-N
Synonym 3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
PubChem CID 11428
ChEBI CHEBI:77519
IUPAC Name pentan-3-ol
SMILES CCC(CC)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0606522
View Documents
Thermo Scientific Chemicals
L0606522
100 g N/A
 
AAL0606536
View Documents
Thermo Scientific Chemicals
L0606536
500 g N/A
 
Description

Description

3-Pentanol is widely used as a flavoring agent. It is biogenic oxygenated volatile organic compound (BOVOC) and is used as a reagent in the synthesis of pure bromopentanes for infrared standards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pentanol is widely used as a flavoring agent. It is biogenic oxygenated volatile organic compound (BOVOC) and is used as a reagent in the synthesis of pure bromopentanes for infrared standards.

Solubility
Soluble in acetone, benzene, ethanol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

584-02-1
88.15
AQIXEPGDORPWBJ-UHFFFAOYSA-N
11428
pentan-3-ol
C5H12O
MFCD00004574
3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
CHEBI:77519
CCC(CC)O

Specifications

584-02-1
0.817
33°C (91°F)
C5H12O
MFCD00004574
UN1105
14,7120
Soluble in acetone,benzene,ethanol and diethyl ether.
CCC(CC)O
88.15
CHEBI:77519
≥98%
-8°C
114°C to 115°C
Characteristic
1.41
100 g
1730964
3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
AQIXEPGDORPWBJ-UHFFFAOYSA-N
pentan-3-ol
11428
88.15
3-Pentanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H315-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P380-P501c

H226-H302-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS

EINECSNumber : 209-526-7

RTECSNumber : SA5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only