missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Pentanone 98.0+%, TCI America™
$41.48 - $112.57
Chemical Identifiers
CAS | 96-22-0 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.13 |
MDL Number | MFCD00009320 |
InChI Key | FDPIMTJIUBPUKL-UHFFFAOYSA-N |
Synonym | 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone |
PubChem CID | 7288 |
ChEBI | CHEBI:67886 |
IUPAC Name | pentan-3-one |
SMILES | CCC(=O)CC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
P006125ML
|
TCI America
P006125ML |
25 mL |
Each for $41.48
|
|
||||||
P0061500ML
|
TCI America
P0061500ML |
500 mL |
Each for $112.57
|
|
||||||
Chemical Identifiers
96-22-0 | |
86.13 | |
FDPIMTJIUBPUKL-UHFFFAOYSA-N | |
7288 | |
pentan-3-one |
C5H10O | |
MFCD00009320 | |
3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone | |
CHEBI:67886 | |
CCC(=O)CC |
Specifications
96-22-0 | |
102°C | |
MFCD00009320 | |
1156 | |
FDPIMTJIUBPUKL-UHFFFAOYSA-N | |
pentan-3-one | |
7288 | |
86.13 | |
Liquid |
Colorless | |
C5H10O | |
25 mL | |
3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone | |
CCC(=O)CC | |
86.13 | |
CHEBI:67886 | |
≥98.0% (GC) | |
3-Pentanone |
Safety and Handling
EINECSNumber : (2)-2474
RTECSNumber : SA8050000
TSCA : Yes