missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC63601DE
Specifications
3-(phenoxymethyl)benzoic acid | |
97% | |
C14H12O3 | |
3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg | |
C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O | |
228.247 | |
228.25 |
31719-75-2 | |
Amber Glass Bottle | |
5g | |
URLYREZIPSJJQU-UHFFFAOYSA-N | |
3-(phenoxymethyl)benzoic acid | |
3729884 | |
97% |
Chemical Identifiers
31719-75-2 | |
228.247 | |
3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg | |
3-(phenoxymethyl)benzoic acid |
C14H12O3 | |
URLYREZIPSJJQU-UHFFFAOYSA-N | |
3729884 | |
C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O |
Safety and Handling
GHS Signal Word: Warning