missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxypiperidine, 97%

CAS: 151666-08-9 | C11H15NO | 177.25 g/mol

$154.47 - $522.53

Chemical Identifiers

CAS 151666-08-9
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD04117746
InChI Key QDPNIJKUTWDGMV-UHFFFAOYNA-N
Synonym piperidine, 3-phenoxy, 3-phenoxy-piperidine, piperidine,3-phenoxy, 3-phenoxypiperidine,, 3-phenoxy piperidine, acmc-20a6se
PubChem CID 3730581
IUPAC Name 3-phenoxypiperidine
SMILES C1CNCC(C1)OC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50912MD
View Documents
Thermo Scientific Chemicals
H50912MD
250 mg N/A
Only null left
 
AAH5091203
View Documents
Thermo Scientific Chemicals
H5091203
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

151666-08-9
177.25
QDPNIJKUTWDGMV-UHFFFAOYNA-N
3730581
C1CNCC(C1)OC1=CC=CC=C1
C11H15NO
MFCD04117746
piperidine, 3-phenoxy, 3-phenoxy-piperidine, piperidine,3-phenoxy, 3-phenoxypiperidine,, 3-phenoxy piperidine, acmc-20a6se
3-phenoxypiperidine

Specifications

151666-08-9
1.543
250 mg
QDPNIJKUTWDGMV-UHFFFAOYNA-N
3-phenoxypiperidine
3730581
97%
C11H15NO
MFCD04117746
piperidine, 3-phenoxy, 3-phenoxy-piperidine, piperidine,3-phenoxy, 3-phenoxypiperidine,, 3-phenoxy piperidine, acmc-20a6se
C1CNCC(C1)OC1=CC=CC=C1
177.25
177.25
3-Phenoxypiperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only