missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals

$149.21 - $514.99

Chemical Identifiers

CAS 136-40-3
Molecular Formula C11H12ClN5
Molecular Weight (g/mol) 249.702
MDL Number MFCD00035347
InChI Key QQBPIHBUCMDKFG-UHFFFAOYSA-N
Synonym phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine
PubChem CID 8691
ChEBI CHEBI:71419
IUPAC Name 3-phenyldiazenylpyridine-2,6-diamine;hydrochloride
SMILES C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2188606
View Documents
Thermo Scientific Chemicals
B2188606
5 g
Each for $149.21
 
AAB2188614
View Documents
Thermo Scientific Chemicals
B2188614
25 g
Each for $514.99
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

136-40-3
249.702
QQBPIHBUCMDKFG-UHFFFAOYSA-N
8691
3-phenyldiazenylpyridine-2,6-diamine;hydrochloride
C11H12ClN5
MFCD00035347
phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine
CHEBI:71419
C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl

Specifications

136-40-3
C11H12ClN5
5 g
QQBPIHBUCMDKFG-UHFFFAOYSA-N
3-phenyldiazenylpyridine-2,6-diamine;hydrochloride
8691
249.7
3-Phenylazo-2,6-diaminopyridine hydrochloride
∼240°C (decomposition)
MFCD00035347
phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine
C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl
249.702
CHEBI:71419
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H351-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H335-H350

EINECSNumber : 205-243-8

RTECSNumber : US7875000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only