missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-3-Phenyllactic acid, 98+%, Thermo Scientific Chemicals

$63.74 - $197.23

Chemical Identifiers

CAS 828-01-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00065928
InChI Key VOXXWSYKYCBWHO-UHFFFAOYSA-N
Synonym dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
PubChem CID 3848
ChEBI CHEBI:25998
IUPAC Name 2-hydroxy-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0985403
View Documents
Thermo Scientific Chemicals
L0985403
1 g
Each for $63.74
 
AAL0985406
View Documents
Thermo Scientific Chemicals
L0985406
5 g
Each for $197.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

828-01-3
166.176
VOXXWSYKYCBWHO-UHFFFAOYSA-N
3848
2-hydroxy-3-phenylpropanoic acid
C9H10O3
MFCD00065928
dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
CHEBI:25998
C1=CC=C(C=C1)CC(C(=O)O)O

Specifications

828-01-3
Odorless
MFCD00065928
2209791
VOXXWSYKYCBWHO-UHFFFAOYSA-N
2-hydroxy-3-phenylpropanoic acid
3848
166.18
(+/-)-3-Phenyllactic acid
95°C to 98°C
C9H10O3
1 g
dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
C1=CC=C(C=C1)CC(C(=O)O)O
166.176
CHEBI:25998
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P305+P351+P338
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 212-580-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only