missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrazolidinone hydrochloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 1752-88-1
Molecular Formula C3H7ClN2O
Molecular Weight (g/mol) 122.55
MDL Number MFCD00043787
InChI Key HAYWTXQRYXYRAF-UHFFFAOYSA-N
Synonym 3-pyrazolidinone hydrochloride, pyrazolidin-3-one hydrochloride, 4,5-dihydro-1h-pyrazol-3-ol hydrochloride, 3-pyrazolidinone, monohydrochloride, tetrahydro-3h-pyrazol-3-one hydrochloride, 3-pyrazolidinone hcl, 3-pyrazolidone hydrochloride, ksc493k0l, pyrazolidin-3-on-hydrochloride, pyrazolidin-3-one hydrogen chloride
PubChem CID 2733712
IUPAC Name pyrazolidin-3-one;hydrochloride
SMILES [H+].[Cl-].O=C1CCNN1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC335490010
SDS View Documents
Thermo Scientific Chemicals
335490010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC335490050
SDS View Documents
Thermo Scientific Chemicals
335490050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC335490250
SDS View Documents
Thermo Scientific Chemicals
335490250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1752-88-1
122.55
HAYWTXQRYXYRAF-UHFFFAOYSA-N
2733712
[H+].[Cl-].O=C1CCNN1
C3H7ClN2O
MFCD00043787
3-pyrazolidinone hydrochloride, pyrazolidin-3-one hydrochloride, 4,5-dihydro-1h-pyrazol-3-ol hydrochloride, 3-pyrazolidinone, monohydrochloride, tetrahydro-3h-pyrazol-3-one hydrochloride, 3-pyrazolidinone hcl, 3-pyrazolidone hydrochloride, ksc493k0l, pyrazolidin-3-on-hydrochloride, pyrazolidin-3-one hydrogen chloride
pyrazolidin-3-one;hydrochloride

Specifications

1752-88-1
98%
C3H7ClN2O
1g
Solubility in water: soluble
[H+].[Cl-].O=C1CCNN1
122.55
122.55
3-Pyrazolidinone hydrochloride
Authentic
Glass bottle
MFCD00043787
3-pyrazolidinone hydrochloride, pyrazolidin-3-one hydrochloride, 4,5-dihydro-1h-pyrazol-3-ol hydrochloride, 3-pyrazolidinone, monohydrochloride, tetrahydro-3h-pyrazol-3-one hydrochloride, 3-pyrazolidinone hcl, 3-pyrazolidone hydrochloride, ksc493k0l, pyrazolidin-3-on-hydrochloride, pyrazolidin-3-one hydrogen chloride
HAYWTXQRYXYRAF-UHFFFAOYSA-N
pyrazolidin-3-one;hydrochloride
2733712
98%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only