missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrid-2-ylbenzaldehyde, ≥97%, Thermo Scientific™

$123.47 - $123.47

Chemical Identifiers

CAS 85553-53-3
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD02684103
InChI Key SAPNGHSAYQXRPG-UHFFFAOYSA-N
Synonym 3-2-pyridyl benzaldehyde, 3-pyridin-2-yl benzaldehyde, 3-2-pyridinyl benzaldehyde, 3-pyridin-2-yl-benzaldehyde, 3-pyrid-2-ylbenzaldehyde, benzaldehyde, 3-2-pyridinyl, 2-3-formylphenyl pyridine, acmc-209q6q, benzaldehyde,3-2-pyridinyl, 5-2-pyridyl benzaldehyde
PubChem CID 3710039
IUPAC Name 3-pyridin-2-ylbenzaldehyde
SMILES C1=CC=NC(=C1)C2=CC(=CC=C2)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC41604DA
SDS View Documents
Thermo Scientific Chemicals
CC41604DA
1 g
Each for $123.47
Only null left
 
Description

Description

Specifications

Chemical Identifiers

85553-53-3
183.21
SAPNGHSAYQXRPG-UHFFFAOYSA-N
3710039
C1=CC=NC(=C1)C2=CC(=CC=C2)C=O
C12H9NO
MFCD02684103
3-2-pyridyl benzaldehyde, 3-pyridin-2-yl benzaldehyde, 3-2-pyridinyl benzaldehyde, 3-pyridin-2-yl-benzaldehyde, 3-pyrid-2-ylbenzaldehyde, benzaldehyde, 3-2-pyridinyl, 2-3-formylphenyl pyridine, acmc-209q6q, benzaldehyde,3-2-pyridinyl, 5-2-pyridyl benzaldehyde
3-pyridin-2-ylbenzaldehyde

Specifications

85553-53-3
100.0
Amber Glass Bottle
MFCD02684103
3-2-pyridyl benzaldehyde, 3-pyridin-2-yl benzaldehyde, 3-2-pyridinyl benzaldehyde, 3-pyridin-2-yl-benzaldehyde, 3-pyrid-2-ylbenzaldehyde, benzaldehyde, 3-2-pyridinyl, 2-3-formylphenyl pyridine, acmc-209q6q, benzaldehyde,3-2-pyridinyl, 5-2-pyridyl benzaldehyde
C1=CC=NC(=C1)C2=CC(=CC=C2)C=O
183.21
183.21
3-Pyrid-2-ylbenzaldehyde
96.5
97%
C12H9NO
1 g
SAPNGHSAYQXRPG-UHFFFAOYSA-N
3-pyridin-2-ylbenzaldehyde
3710039
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'