Learn More
3-Pyridinesulfonic acid, 98%
CAS: 636-73-7 | C5H4NO3S | 158.15 g/mol
$236.06 - $236.06
Chemical Identifiers
| CAS | 636-73-7 |
|---|---|
| Molecular Formula | C5H4NO3S |
| Molecular Weight (g/mol) | 158.15 |
| MDL Number | MFCD00006381 |
| InChI Key | DVECLMOWYVDJRM-UHFFFAOYSA-M |
| Synonym | 3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7 |
| PubChem CID | 69468 |
| IUPAC Name | pyridine-3-sulfonic acid |
| SMILES | [O-]S(=O)(=O)C1=CC=CN=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC131921000
|
Thermo Scientific Chemicals
131921000 |
100 g | Plastic bottle |
Each for $236.06
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 636-73-7 | |
| 158.15 | |
| DVECLMOWYVDJRM-UHFFFAOYSA-M | |
| 69468 | |
| [O-]S(=O)(=O)C1=CC=CN=C1 |
| C5H4NO3S | |
| MFCD00006381 | |
| 3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7 | |
| pyridine-3-sulfonic acid |
Specifications
| 636-73-7 | |
| 100.0 | |
| White | |
| Authentic | |
| Plastic bottle | |
| (C5H4N)SO3H | |
| 100 g | |
| 3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7 | |
| DVECLMOWYVDJRM-UHFFFAOYSA-M | |
| pyridine-3-sulfonic acid | |
| 69468 | |
| 98% | |
| 3-Pyridinesulfonic acid, 98% |
| 97.5 | |
| >300.0°C | |
| >66°C | |
| 98% | |
| C5H4NO3S | |
| MFCD00006381 | |
| 22, 387 | |
| (5% in water) Clear colorless | |
| [O-]S(=O)(=O)C1=CC=CN=C1 | |
| 158.15 | |
| 159.16 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 211-265-9
RTECSNumber : UT7920000
TSCA : TSCA
RUO – Research Use Only