missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pyridyl isothiocyanate, ≥95%, Thermo Scientific™

$85.67 - $85.67

Chemical Identifiers

CAS 17452-27-6
Molecular Formula C6H4N2S
Molecular Weight (g/mol) 136.172
MDL Number MFCD00052351
InChI Key VMSZFBSYWXMXRF-UHFFFAOYSA-N
Synonym 3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
PubChem CID 2737242
IUPAC Name 3-isothiocyanatopyridine
SMILES C1=CC(=CN=C1)N=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC04107DA
SDS View Documents
Thermo Scientific Chemicals
CC04107DA
1 g Amber Glass Bottle
Each for $85.67
 
Description

Description

Specifications

Chemical Identifiers

17452-27-6
136.172
VMSZFBSYWXMXRF-UHFFFAOYSA-N
2737242
C1=CC(=CN=C1)N=C=S
C6H4N2S
MFCD00052351
3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
3-isothiocyanatopyridine

Specifications

17452-27-6
100.0
95%
C6H4N2S
1 g
VMSZFBSYWXMXRF-UHFFFAOYSA-N
3-isothiocyanatopyridine
2737242
95%
94.5
1.22g/mL
Amber Glass Bottle
MFCD00052351
3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
C1=CC(=CN=C1)N=C=S
136.172
136.18
3-Pyridyl isothiocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'