Learn More
3-Pyridylacetic acid hydrochloride, 98%, Thermo Scientific Chemicals
$127.64 - $747.29
Chemical Identifiers
CAS | 6419-36-9 |
---|---|
Molecular Formula | C7H7NO2·ClH |
Molecular Weight (g/mol) | 173.6 |
MDL Number | MFCD00012819 |
InChI Key | XVCCOEWNFXXUEV-UHFFFAOYSA-N |
Synonym | 3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride |
PubChem CID | 2723724 |
IUPAC Name | 2-pyridin-3-ylacetic acid;hydrochloride |
SMILES | C1=CC(=CN=C1)CC(=O)O.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC131960025
|
Thermo Scientific Chemicals
131960025 |
2.5 g | Glass bottle |
Each for $127.64
|
|
||||
AC131960250
|
Thermo Scientific Chemicals
131960250 |
25 g | Glass bottle |
Each for $747.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6419-36-9 | |
173.6 | |
XVCCOEWNFXXUEV-UHFFFAOYSA-N | |
2723724 | |
C1=CC(=CN=C1)CC(=O)O.Cl |
C7H7NO2·ClH | |
MFCD00012819 | |
3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride | |
2-pyridin-3-ylacetic acid;hydrochloride |
Specifications
6419-36-9 | |
100.0 | |
Beige or White to Yellow | |
97.5% min. (Argentometry) | |
C7H7NO2·ClH | |
MFCD00012819 | |
10, 4635 | |
Solubility in water: soluble | |
C1=CC(=CN=C1)CC(=O)O.Cl | |
173.6 | |
173.6 | |
Crystalline Powder, Crystals or Chunks |
97.5 | |
160°C to 161°C | |
Authentic | |
Glass bottle | |
(C5H4N)CH2OOH·HCl | |
2.5 g | |
3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride | |
XVCCOEWNFXXUEV-UHFFFAOYSA-N | |
2-pyridin-3-ylacetic acid;hydrochloride | |
2723724 | |
98% | |
3-Pyridylacetic acid hydrochloride |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 229-148-6
RUO – Research Use Only