missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(tert-Butyldimethylsilyloxy)phenylboronic acid, 96+%, Thermo Scientific™

Chemical Identifiers

CAS 261621-12-9
Molecular Formula C12H21BO3Si
Molecular Weight (g/mol) 252.19
MDL Number MFCD03701505
InChI Key RDQWADDNQONTLB-UHFFFAOYSA-N
Synonym 3-tert-butyldimethylsilyl oxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy phenylboronic acid, 3-t-butyldimethylsilyloxy phenylboronic acid, 3-tert-butyl dimethylsiloxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy benzeneboronic acid, 3-tert-butyldimethylsiloxy benzeneboronic acid, 3-tert-butyldimethylsilyl oxy phenylboronic acid, 3-tert-butyl dimethyl silyl oxyphenyl boronic acid, m-t-butyldimethylsilyloxy phenylboronic acid, 3-t-butyl dimethylsiloxy phenyl boronic acid
PubChem CID 3463029
IUPAC Name [3-[tert-butyl(dimethyl)silyl]oxyphenyl]boronic acid
SMILES B(C1=CC(=CC=C1)O[Si](C)(C)C(C)(C)C)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398340010
SDS View Documents
Thermo Scientific Chemicals
398340010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398340050
SDS View Documents
Thermo Scientific Chemicals
398340050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

261621-12-9
252.19
RDQWADDNQONTLB-UHFFFAOYSA-N
3463029
B(C1=CC(=CC=C1)O[Si](C)(C)C(C)(C)C)(O)O
C12H21BO3Si
MFCD03701505
3-tert-butyldimethylsilyl oxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy phenylboronic acid, 3-t-butyldimethylsilyloxy phenylboronic acid, 3-tert-butyl dimethylsiloxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy benzeneboronic acid, 3-tert-butyldimethylsiloxy benzeneboronic acid, 3-tert-butyldimethylsilyl oxy phenylboronic acid, 3-tert-butyl dimethyl silyl oxyphenyl boronic acid, m-t-butyldimethylsilyloxy phenylboronic acid, 3-t-butyl dimethylsiloxy phenyl boronic acid
[3-[tert-butyl(dimethyl)silyl]oxyphenyl]boronic acid

Specifications

261621-12-9
100.0
96+%
C12H21BO3Si
MFCD03701505
3-tert-butyldimethylsilyl oxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy phenylboronic acid, 3-t-butyldimethylsilyloxy phenylboronic acid, 3-tert-butyl dimethylsiloxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy benzeneboronic acid, 3-tert-butyldimethylsiloxy benzeneboronic acid, 3-tert-butyldimethylsilyl oxy phenylboronic acid, 3-tert-butyl dimethyl silyl oxyphenyl boronic acid, m-t-butyldimethylsilyloxy phenylboronic acid, 3-t-butyl dimethylsiloxy phenyl boronic acid
B(C1=CC(=CC=C1)O[Si](C)(C)C(C)(C)C)(O)O
252.19
252.19
3-(tert-Butyldimethylsilyloxy)phenylboronic acid
96.0
Authentic
Glass bottle
(CH3)CSi(CH3)2OC6H4B(OH)2
1g
RDQWADDNQONTLB-UHFFFAOYSA-N
[3-[tert-butyl(dimethyl)silyl]oxyphenyl]boronic acid
3463029
96+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only