missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-tert-Butylphenol, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 154381000

Catalog No. AC154381000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-tert-Butylphenol
585-34-2
100.0
108°C
99% min. (GC)
C10H14O
MFCD00002300
m-tert-butylphenol, phenol, 3-1,1-dimethylethyl, 3-t-butylphenol, 3-tert-butyl phenol, phenol, m-tert-butyl, 3-tert-butyl-phenol, 3-tertbutylphenol, 3-tert.butylphenol, m-tert.-butylphenol
CYEKUDPFXBLGHH-UHFFFAOYSA-N
3-tert-butylphenol
11450
150.22
99+%
99.0
125°C to 130°C (20.0mmHg)
Authentic
Plastic bottle
(CH3)3CC6H4OH
100g
Solubility in water: .. Other solubilities: soluble in methanol
CC(C)(C)C1=CC=CC(O)=C1
150.22
CHEBI:34348
99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-553-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only