missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Thien-2-ylbenzonitrile 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC44516DA
Specifications
3-Thien-2-ylbenzonitrile | |
97% | |
C11H7NS | |
1g | |
DIILPQRSOQWSCU-UHFFFAOYSA-N | |
3-thiophen-2-ylbenzonitrile | |
4645033 | |
97% |
380626-35-7 | |
Amber Glass Bottle | |
MFCD02323389 | |
3-thien-2-ylbenzonitrile, 3-thiophen-2-yl benzonitrile, 3-2-thienyl benzonitrile, 3-thiophen-2-yl-benzonitrile, 3-2-thienyl benzenecarbonitrile | |
N#CC1=CC(=CC=C1)C1=CC=CS1 | |
185.24 | |
185.25 |
Chemical Identifiers
380626-35-7 | |
185.24 | |
DIILPQRSOQWSCU-UHFFFAOYSA-N | |
4645033 | |
N#CC1=CC(=CC=C1)C1=CC=CS1 |
C11H7NS | |
MFCD02323389 | |
3-thien-2-ylbenzonitrile, 3-thiophen-2-yl benzonitrile, 3-2-thienyl benzonitrile, 3-thiophen-2-yl-benzonitrile, 3-2-thienyl benzenecarbonitrile | |
3-thiophen-2-ylbenzonitrile |