missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Trifluoromethoxy)benzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America T23341G
Specifications
3-(Trifluoromethoxy)benzoyl Chloride | |
Colorless | |
MFCD00061268 | |
3265 | |
MGKIOAXLPYJSMU-UHFFFAOYSA-N | |
3-(trifluoromethoxy)benzoyl chloride | |
522854 | |
≥98.0% (GC) |
86270-03-3 | |
C8H4ClF3O2 | |
1 g | |
3-trifluoromethoxy benzoyl chloride, m-trifluoromethoxybenzoyl chloride, m-trifluoromethoxy benzoyl chloride, benzoyl chloride, 3-trifluoromethoxy, 3-trifluoromethoxy-benzoyl chloride, benzoylchloride, 3-trifluoromethoxy, ksc450s6d, m-trifluoromethoxybenzoylchloride, 3-trifluoromethoxy benzoylchloride, 3-trifluoromethyloxy benzoyl chloride | |
C1=CC(=CC(=C1)OC(F)(F)F)C(=O)Cl | |
224.563 | |
224.56 | |
Liquid |
Chemical Identifiers
86270-03-3 | |
224.563 | |
MGKIOAXLPYJSMU-UHFFFAOYSA-N | |
522854 | |
C1=CC(=CC(=C1)OC(F)(F)F)C(=O)Cl |
C8H4ClF3O2 | |
MFCD00061268 | |
3-trifluoromethoxy benzoyl chloride, m-trifluoromethoxybenzoyl chloride, m-trifluoromethoxy benzoyl chloride, benzoyl chloride, 3-trifluoromethoxy, 3-trifluoromethoxy-benzoyl chloride, benzoylchloride, 3-trifluoromethoxy, ksc450s6d, m-trifluoromethoxybenzoylchloride, 3-trifluoromethoxy benzoylchloride, 3-trifluoromethyloxy benzoyl chloride | |
3-(trifluoromethoxy)benzoyl chloride |
Safety and Handling
TSCA : No