missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)benzyl mercaptan, tech. 90%, Thermo Scientific Chemicals

$96.56 - $312.74

Chemical Identifiers

CAS 25697-55-6
Molecular Formula C8H7F3S
Molecular Weight (g/mol) 192.199
MDL Number MFCD00042435
InChI Key CQIQWIMXCPTQPJ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl mercaptan, 3-trifluoromethyl phenyl methanethiol, 3-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl phenylmethanethiol, 3-trifluoromethyl benzylthiol, m-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl benzylmercaptan, 3-trifluoromethyl phenyl methane-1-thiol, acmc-1cnzu
PubChem CID 141236
IUPAC Name [3-(trifluoromethyl)phenyl]methanethiol
SMILES C1=CC(=CC(=C1)C(F)(F)F)CS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0815503
View Documents
Thermo Scientific Chemicals
L0815503
1 g
Each for $96.56
 
AAL0815506
View Documents
Thermo Scientific Chemicals
L0815506
5 g
Each for $312.74
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25697-55-6
192.199
CQIQWIMXCPTQPJ-UHFFFAOYSA-N
141236
C1=CC(=CC(=C1)C(F)(F)F)CS
C8H7F3S
MFCD00042435
3-trifluoromethyl benzyl mercaptan, 3-trifluoromethyl phenyl methanethiol, 3-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl phenylmethanethiol, 3-trifluoromethyl benzylthiol, m-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl benzylmercaptan, 3-trifluoromethyl phenyl methane-1-thiol, acmc-1cnzu
[3-(trifluoromethyl)phenyl]methanethiol

Specifications

25697-55-6
C8H7F3S
MFCD00042435
UN2810
Air Sensitive
CQIQWIMXCPTQPJ-UHFFFAOYSA-N
[3-(trifluoromethyl)phenyl]methanethiol
141236
90%
3-(Trifluoromethyl)benzyl mercaptan
75°C to 77°C (5 mmHg)
1.49
1 g
2520097
3-trifluoromethyl benzyl mercaptan, 3-trifluoromethyl phenyl methanethiol, 3-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl phenylmethanethiol, 3-trifluoromethyl benzylthiol, m-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl benzylmercaptan, 3-trifluoromethyl phenyl methane-1-thiol, acmc-1cnzu
C1=CC(=CC(=C1)C(F)(F)F)CS
192.199
192.2
Technical
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only