missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Trifluoromethyl)phenylacetic acid, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1096009
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-(Trifluoromethyl)phenylacetic acid | |
77°C to 78°C | |
MFCD00004339 | |
2213223 | |
BLXXCCIBGGBDHI-UHFFFAOYSA-N | |
2-[3-(trifluoromethyl)phenyl]acetic acid | |
67695 | |
≥98% |
351-35-9 | |
C9H7F3O2 | |
10 g | |
3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride | |
C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O | |
204.148 | |
204.15 |
Chemical Identifiers
351-35-9 | |
204.148 | |
BLXXCCIBGGBDHI-UHFFFAOYSA-N | |
67695 | |
C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O |
C9H7F3O2 | |
MFCD00004339 | |
3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride | |
2-[3-(trifluoromethyl)phenyl]acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-511-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only