Learn More
3-(Trifluoromethyl)phenylhydrazine, 95%
CAS: 368-78-5 | C7H7F3N2 | 176.142 g/mol
$93.40 - $285.99
Chemical Identifiers
| CAS | 368-78-5 |
|---|---|
| Molecular Formula | C7H7F3N2 |
| Molecular Weight (g/mol) | 176.142 |
| MDL Number | MFCD00025093 |
| InChI Key | RSESUCWJKLHXEZ-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethyl phenylhydrazine, 3-trifluoromethyl phenyl hydrazine, 3-trifluoromethylphenylhydrazine, hydrazine, 3-trifluoromethyl phenyl, 3-trifloromethylphenylhydrazine, 3-trifluouromethyl phenylhydrazinehydrochloride, 3-trifluoromethyl phenylhydrazine hydrochioride, pubchem3280, enamine_001261 |
| PubChem CID | 98650 |
| IUPAC Name | [3-(trifluoromethyl)phenyl]hydrazine |
| SMILES | C1=CC(=CC(=C1)NN)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1199603
|
Thermo Scientific Chemicals
L1199603 |
1 g |
Each for $93.40
|
|
|||||
|
AAL1199606
|
Thermo Scientific Chemicals
L1199606 |
5 g |
Each for $285.99
|
|
|||||
Description
3-(Trifluoromethyl)phenylhydrazine is used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Trifluoromethyl)phenylhydrazine is used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 368-78-5 | |
| 176.142 | |
| RSESUCWJKLHXEZ-UHFFFAOYSA-N | |
| 98650 | |
| C1=CC(=CC(=C1)NN)C(F)(F)F |
| C7H7F3N2 | |
| MFCD00025093 | |
| 3-trifluoromethyl phenylhydrazine, 3-trifluoromethyl phenyl hydrazine, 3-trifluoromethylphenylhydrazine, hydrazine, 3-trifluoromethyl phenyl, 3-trifloromethylphenylhydrazine, 3-trifluouromethyl phenylhydrazinehydrochloride, 3-trifluoromethyl phenylhydrazine hydrochioride, pubchem3280, enamine_001261 | |
| [3-(trifluoromethyl)phenyl]hydrazine |
Specifications
| 368-78-5 | |
| 80°C to 83°C (9 mmHg) | |
| C7H7F3N2 | |
| MFCD00025093 | |
| 640075 | |
| Not miscible or difficult to mix with water. | |
| C1=CC(=CC(=C1)NN)C(F)(F)F | |
| 176.142 | |
| 176.14 | |
| 3-(Trifluoromethyl)phenylhydrazine |
| 1.348 | |
| 107°C (224°F) | |
| 1.504 | |
| 1 g | |
| 3-trifluoromethyl phenylhydrazine, 3-trifluoromethyl phenyl hydrazine, 3-trifluoromethylphenylhydrazine, hydrazine, 3-trifluoromethyl phenyl, 3-trifloromethylphenylhydrazine, 3-trifluouromethyl phenylhydrazinehydrochloride, 3-trifluoromethyl phenylhydrazine hydrochioride, pubchem3280, enamine_001261 | |
| RSESUCWJKLHXEZ-UHFFFAOYSA-N | |
| [3-(trifluoromethyl)phenyl]hydrazine | |
| 98650 | |
| 95% |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 206-713-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only