missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)piperidine, 97%

CAS: 768-31-0 | C6H11F3N | 154.16 g/mol

$144.04 - $370.71

Chemical Identifiers

CAS 768-31-0
Molecular Formula C6H11F3N
Molecular Weight (g/mol) 154.16
MDL Number MFCD02183561
InChI Key JOHFJTBDUSVGQB-RXMQYKEDSA-O
PubChem CID 2760776
SMILES FC(F)(F)[C@@H]1CCC[NH2+]C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH25851MD
View Documents
Thermo Scientific Chemicals
H25851MD
250 mg
Each for $144.04
Only null left
 
AAH2585103
View Documents
Thermo Scientific Chemicals
H2585103
1 g
Each for $370.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

768-31-0
154.16
JOHFJTBDUSVGQB-RXMQYKEDSA-O
FC(F)(F)[C@@H]1CCC[NH2+]C1
C6H11F3N
MFCD02183561
2760776

Specifications

768-31-0
128°C to 130°C
Amine-like
1.393
250 mg
Air Sensitive
JOHFJTBDUSVGQB-RXMQYKEDSA-O
(3R)-3-(trifluoromethyl)piperidin-1-ium
2760776
97%
1.16
28°C (83°F)
C6H11F3N
MFCD02183561
UN2733
3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl
FC(F)(F)[C@@H]1CCC[NH2+]C1
154.16
153.15
3-(Trifluoromethyl)piperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H301+H311+H331-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only